Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
170564-98-4
C9H13NO
151.21
MFCD01311791
SEQXIQNPMQTBGN-SECBINFHSA-N
r-1-phenyl-3-propanolamine,r-3-amino-3-phenylpropan-1-ol,3r-3-amino-3-phenylpropan-1-ol,r-beta-phenylalaninol,r-3-amino-3-phenyl-1-propanol,r-3-phenyl-beta-alaninol,r-3-amino-3-phenyl-propan-1-ol,r--phenylalaninol,pubchem13853
7016858
(3R)-3-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)C(CCO)N
170564-98-4
C9H13NO
151.21
MFCD01311791
SEQXIQNPMQTBGN-SECBINFHSA-N
r-1-phenyl-3-propanolamine,r-3-amino-3-phenylpropan-1-ol,3r-3-amino-3-phenylpropan-1-ol,r-beta-phenylalaninol,r-3-amino-3-phenyl-1-propanol,r-3-phenyl-beta-alaninol,r-3-amino-3-phenyl-propan-1-ol,r--phenylalaninol,pubchem13853
7016858
(3R)-3-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)C(CCO)N
Infrared Spectrum Authentic
Packaging Glass bottle
Enantiomeric Excess 97.5% min.
Color White to Yellow or Yellow
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 151.21
Physical Form Viscous Liquid or Low Melting Solid
Percent Purity 95%,98% ee
Chemical Name or Material (R)-3-Amino-3-phenylpropan-1-ol, 95%, (98% e.e.)
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA