Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-3-Amino-3-phenylpropionic acid hydrochloride, ≥98%, Acros Organics

Click to view available options
Quantity:
50mg
83649-48-3
C9H12ClNO2
201.65
MFCD03092946
ABEBCTCOPRULFS-DDWIOCJRSA-N
r-3-amino-3-phenylpropanoic acid hydrochloride,r-3-amino-3-phenylpropionic acid hydrochloride,3r-3-amino-3-phenylpropanoic acid hydrochloride,l-beta-homophenylglycine hydrochloride,r-3-amino-3-phenylpropionic acid hcl,pubchem5925,d-beta-phenylalanine hcl,r-beta-homophenylglycine hydrochloride,3r-3-amino-3-phenylpropanoic acid, chloride,r-3-amino-3-phenyl-propionic acid hcl salt
2734697
(3R)-3-amino-3-phenylpropanoic acid;hydrochloride
C1=CC=C(C=C1)C(CC(=O)O)N.Cl
83649-48-3
C9H12ClNO2
201.65
MFCD03092946
ABEBCTCOPRULFS-DDWIOCJRSA-N
r-3-amino-3-phenylpropanoic acid hydrochloride,r-3-amino-3-phenylpropionic acid hydrochloride,3r-3-amino-3-phenylpropanoic acid hydrochloride,l-beta-homophenylglycine hydrochloride,r-3-amino-3-phenylpropionic acid hcl,pubchem5925,d-beta-phenylalanine hcl,r-beta-homophenylglycine hydrochloride,3r-3-amino-3-phenylpropanoic acid, chloride,r-3-amino-3-phenyl-propionic acid hcl salt
2734697
(3R)-3-amino-3-phenylpropanoic acid;hydrochloride
C1=CC=C(C=C1)C(CC(=O)O)N.Cl
Quantity 50mg
Infrared Spectrum Authentic
Color Beige
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 201.66
Physical Form Powder
Percent Purity ≥98.0%
Chemical Name or Material (R)-(-)-3-Amino-3-phenylpropionic acid hydrochloride