Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
111769-27-8
C7H8O3S·C4H9NO
259.32
BZXPLADBSZWDIH-FZSMXKCYSA-N
r-3-aminotetrahydrofuran tosylate,r-+-tetrahydro-3-furylamine p-toluenesulfonate salt,r---3-aminotetrahydrofuran toluene-4-sulfonate,r-+-3-aminotetrahydrofuran toluene-4-sulfonate,r---3-aminotetrahydrofurantoluene-4-sulfonate,3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate,c4h9no.c7h8o3s,r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate,3r-oxolan-3-amine; para-toluene sulfonate,4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine
14243169
4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine
CC1=CC=C(C=C1)S(=O)(=O)O.C1COCC1N
111769-27-8
C7H8O3S·C4H9NO
259.32
BZXPLADBSZWDIH-FZSMXKCYSA-N
r-3-aminotetrahydrofuran tosylate,r-+-tetrahydro-3-furylamine p-toluenesulfonate salt,r---3-aminotetrahydrofuran toluene-4-sulfonate,r-+-3-aminotetrahydrofuran toluene-4-sulfonate,r---3-aminotetrahydrofurantoluene-4-sulfonate,3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate,c4h9no.c7h8o3s,r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate,3r-oxolan-3-amine; para-toluene sulfonate,4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine
14243169
4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine
CC1=CC=C(C=C1)S(=O)(=O)O.C1COCC1N
Specific Rotation 4.3
Melting Point 137.3°C to 141°C
Color White
Infrared Spectrum Authentic
Quantity 1g
Elemental Analysis 48.91 to 52.99% C, 5.18 to 5.62% N
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 259.32
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material (R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning