Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-(Benzyloxycarbonyl)-4-oxazolidinecarboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
97534-84-4
C12H13NO5
251.238
MFCD00274192
XRRRGBIMHQARMF-SNVBAGLBSA-N
r-+-3-benzyloxycarbonyl-4-oxazolidinecarboxylic acid,r-cbz-oxaproline,r-3-benzyloxy carbonyl oxazolidine-4-carboxylic acid,4r-3-benzyloxycarbonyl oxazolidine-4-carboxylic acid,4r-3-phenylmethoxycarbonyl-1,3-oxazolidine-4-carboxylic acid,r--3-benzyl-oxycarbonyl-4-oxazolidinecarboxylic acid,r-+-3-benzyl-oxycarbonyl-4-oxazolidinecarboxylic acid,3,4-oxazolidinedicarboxylicacid,3-phenylmethyl ester, 4r
688329
(4R)-3-phenylmethoxycarbonyl-1,3-oxazolidine-4-carboxylic acid
C1C(N(CO1)C(=O)OCC2=CC=CC=C2)C(=O)O
97534-84-4
C12H13NO5
251.238
MFCD00274192
XRRRGBIMHQARMF-SNVBAGLBSA-N
r-+-3-benzyloxycarbonyl-4-oxazolidinecarboxylic acid,r-cbz-oxaproline,r-3-benzyloxy carbonyl oxazolidine-4-carboxylic acid,4r-3-benzyloxycarbonyl oxazolidine-4-carboxylic acid,4r-3-phenylmethoxycarbonyl-1,3-oxazolidine-4-carboxylic acid,r--3-benzyl-oxycarbonyl-4-oxazolidinecarboxylic acid,r-+-3-benzyl-oxycarbonyl-4-oxazolidinecarboxylic acid,3,4-oxazolidinedicarboxylicacid,3-phenylmethyl ester, 4r
688329
(4R)-3-phenylmethoxycarbonyl-1,3-oxazolidine-4-carboxylic acid
C1C(N(CO1)C(=O)OCC2=CC=CC=C2)C(=O)O
Specific Rotation +90° to +94° (20°C, 589nm) (C=1, CHCl3)
Quantity 5g
Packaging Glass bottle
Melting Point 76°C to 80°C
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 251.24
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material (R)-(+)-3-(Benzyloxycarbonyl)-4-oxazolidinecarboxylic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.