Alfa Aesar™ (R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid, 98%

Manufacturer: Alfa Aesar™ H2724003

Quantity 1g

 View more versions of this product

Catalog No. AAH2724003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97534-84-4
    Molecular Formula C12H13NO5
    Formula Weight 251.24
    MDL Number MFCD00274192
    Synonym N-Cbz-(R)-oxaproline; Z-D-OxaPro
    Chemical Name or Material (R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid

    Specifications

    Formula Weight 251.24
    Molecular Formula C12H13NO5
    MDL Number MFCD00274192
    CAS 97534-84-4
    Synonym N-Cbz-(R)-oxaproline; Z-D-OxaPro
    Melting Point 80° to 81°C
    Quantity 1g
    Optical Rotation +79° (c=1 in Chloroform)
    Chemical Name or Material (R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit