Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ H2724006

Quantity 5g

 View more versions of this product

Catalog No. AAH2724006


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

97534-84-4
251.24
N-Cbz-(R)-oxaproline; Z-D-OxaPro
C12H13NO5
MFCD00274192
(R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid

Specifications

97534-84-4
251.24
N-Cbz-(R)-oxaproline; Z-D-OxaPro
5g
(R)-(+)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
C12H13NO5
MFCD00274192
80° to 81°C
+79° (c=1 in Chloroform)
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit