Alfa Aesar™ (R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid, 98%

Manufacturer: Alfa Aesar™ H5202806

Quantity 5g

 View more versions of this product

Catalog No. AAH5202806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 500789-07-1
    Molecular Formula C14H18BrNO4
    Formula Weight 344.2
    MDL Number MFCD03427961
    Chemical Name or Material (R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid

    Specifications

    Formula Weight 344.2
    Molecular Formula C14H18BrNO4
    MDL Number MFCD03427961
    CAS 500789-07-1
    Melting Point 142° to 144°C
    Quantity 5g
    Optical Rotation −20° (c=1 in Ethanol)
    Chemical Name or Material (R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit