Alfa Aesar™ (R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid, 98%

Manufacturer: Alfa Aesar™ H5200306

Quantity 5g

 View more versions of this product

Catalog No. AAH5200306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 500789-06-0
    Molecular Formula C14H18ClNO4
    Formula Weight 299.75
    MDL Number MFCD03427959
    Chemical Name or Material (R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid

    Specifications

    Formula Weight 299.75
    Molecular Formula C14H18ClNO4
    MDL Number MFCD03427959
    Optical Rotation +40° (c=1 in Ethanol)
    CAS 500789-06-0
    Melting Point 90° to 92°C
    Quantity 5g
    Chemical Name or Material (R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit