Alfa Aesar™ (R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid, 95%

Manufacturer: Alfa Aesar™ H52166MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52166MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 261380-20-5
    Molecular Formula C14H18BrNO4
    Formula Weight 344.2
    MDL Number MFCD01320860
    Chemical Name or Material (R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid

    Specifications

    Formula Weight 344.2
    Molecular Formula C14H18BrNO4
    MDL Number MFCD01320860
    CAS 261380-20-5
    Melting Point 143° to 145°C
    Quantity 250mg
    Chemical Name or Material (R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit