Alfa Aesar™ (R)-3-(Boc-amino)-4-(4-bromophenyl)butyric acid, 95%

Manufacturer: Alfa Aesar™ H52136MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52136MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 331763-75-8
    Molecular Formula C15H20BrNO4
    Formula Weight 358.23
    MDL Number MFCD01860917
    Chemical Name or Material (R)-3-(Boc-amino)-4-(4-bromophenyl)butyric acid

    Specifications

    Formula Weight 358.23
    Melting Point 144° to 146°C
    Quantity 250mg
    Molecular Formula C15H20BrNO4
    MDL Number MFCD01860917
    CAS 331763-75-8
    Chemical Name or Material (R)-3-(Boc-amino)-4-(4-bromophenyl)butyric acid
    Assay 95%

    Description

    Applications
    (R)-3-(Boc-amino)-4-(4-bromophenyl)butyric acid is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit