Alfa Aesar™ (R)-3-(Boc-amino)-5-phenylpentanoic acid, 95%

Manufacturer: Alfa Aesar™ H5205503

Quantity 1g

 View more versions of this product

Catalog No. AAH5205503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 218608-83-4
    Molecular Formula C16H23NO4
    Formula Weight 293.36
    MDL Number MFCD01076262
    Chemical Name or Material (R)-3-(Boc-amino)-5-phenylpentanoic acid

    Specifications

    Formula Weight 293.36
    Molecular Formula C16H23NO4
    MDL Number MFCD01076262
    CAS 218608-83-4
    Quantity 1g
    Chemical Name or Material (R)-3-(Boc-amino)-5-phenylpentanoic acid
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit