Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(R)-(+)-3-(Boc-amino)pyrrolidine, 99%, ee 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(R)-(+)-3-(Boc-amino)pyrrolidine is used as an intermediate in organic synthesis, as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
(R)-(+)-3-(Boc-amino)pyrrolidine is used as an intermediate in organic synthesis, as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications
Melting Point | 50°C to 54°C |
UN Number | UN3259 |
Beilstein | 5377812 |
Solubility Information | Soluble in water. |
Formula Weight | 186.25 |
Percent Purity | 99% |
Quantity | 1 g |
Optical Rotation | +21° (c=1 in Ethanol) |
Sensitivity | Air sensitive |
Assay | ee 99% |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S. |
TSCA | No |
Recommended Storage | Keep cold |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction