Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Bromo-alpha-methylbenzyl alcohol, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass Ampule with Screw-cap
Glass bottle
134615-24-0
C8H9BrO
201.06
MFCD06658929
ULMJQMDYAOJNCC-ZCFIWIBFSA-N
r-1-3-bromophenyl ethanol,1r-1-3-bromophenyl ethanol,1r-1-3-bromophenyl ethan-1-ol,pubchem15565,r-1-m-bromophenyl ethanol,r-1-3-bromo-phenyl-ethanol,r-3-bromo-alpha-methylbenzyl alcohol,benzenemethanol, 3-bromo-a-methyl-, ar
2734889
(1R)-1-(3-bromophenyl)ethanol
CC(C1=CC(=CC=C1)Br)O
134615-24-0
C8H9BrO
201.06
MFCD06658929
ULMJQMDYAOJNCC-ZCFIWIBFSA-N
r-1-3-bromophenyl ethanol,1r-1-3-bromophenyl ethanol,1r-1-3-bromophenyl ethan-1-ol,pubchem15565,r-1-m-bromophenyl ethanol,r-1-3-bromo-phenyl-ethanol,r-3-bromo-alpha-methylbenzyl alcohol,benzenemethanol, 3-bromo-a-methyl-, ar
2734889
(1R)-1-(3-bromophenyl)ethanol
CC(C1=CC(=CC=C1)Br)O
Packaging Glass bottle
Color Colorless
Enantiomeric Excess 97.5% min. (GC)
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula CH3CH(OH)C6H4Br
Formula Weight 201.06
Physical Form Liquid
Percent Purity 95%,98% ee
Chemical Name or Material (R)-3-Bromo-α-methylbenzyl alcohol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning