Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-chloro-1-phenyl-1-propanol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
100306-33-0
C9H11ClO
170.64
JZFUHAGLMZWKTF-SECBINFHSA-N
r-3-chloro-1-phenylpropan-1-ol,r-+-3-chloro-1-phenyl-1-propanol,r-+-3-chloro-1-phenylpropanol,1r-3-chloro-1-phenylpropan-1-ol,1r-3-chloro-1-phenyl-propan-1-ol,r-3-chloro-1-phenyl-1-propanol,r-3-chloro-1-phenyl-propan-1-ol,r-3-chloro-1-phenylpropanol,pubchem5738
642409
(1R)-3-chloro-1-phenylpropan-1-ol
C1=CC=C(C=C1)C(CCCl)O
100306-33-0
C9H11ClO
170.64
JZFUHAGLMZWKTF-SECBINFHSA-N
r-3-chloro-1-phenylpropan-1-ol,r-+-3-chloro-1-phenyl-1-propanol,r-+-3-chloro-1-phenylpropanol,1r-3-chloro-1-phenylpropan-1-ol,1r-3-chloro-1-phenyl-propan-1-ol,r-3-chloro-1-phenyl-1-propanol,r-3-chloro-1-phenyl-propan-1-ol,r-3-chloro-1-phenylpropanol,pubchem5738
642409
(1R)-3-chloro-1-phenylpropan-1-ol
C1=CC=C(C=C1)C(CCCl)O
Melting Point 59.5°C to 59.8°C
Packaging Glass Bottle
Quantity 5g
Color White to Yellow
Infrared Spectrum Authentic
Specific Rotation +23° to +29° (20°C, 589nm) (c=1 in Chloroform)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 170.64
Physical Form Powder
Percent Purity 97%
Chemical Name or Material (R)-(+)-3-chloro-1-phenyl-1-propanol
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning