Alfa Aesar™ (R)-(+)-3-(Dimethylamino)pyrrolidine, 95%

Manufacturer: Alfa Aesar™ H26251MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26251MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 132958-72-6
    Molecular Formula C6H14N2
    Formula Weight 114.19
    MDL Number MFCD00191347
    UN Number UN2733
    Synonym (3R)-N,N-Dimethyl-3-pyrrolidinamine
    Chemical Name or Material (R)-(+)-3-(Dimethylamino)pyrrolidine

    Specifications

    Formula Weight 114.19
    MDL Number MFCD00191347
    Quantity 250mg
    Molecular Formula C6H14N2
    UN Number UN2733
    Synonym (3R)-N,N-Dimethyl-3-pyrrolidinamine
    Health Hazard 1 Danger
    Boiling Point 166°C
    Flash Point 51°C (124°F)
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    CAS 132958-72-6
    Chemical Name or Material (R)-(+)-3-(Dimethylamino)pyrrolidine
    Assay 95%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    2° to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit