Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-Hydroxypiperidine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
198976-43-1
C5H12ClNO
137.61
MFCD00192221
VLECDMDGMKPUSK-NUBCRITNSA-N
r-3-hydroxypiperidine hydrochloride,r-piperidin-3-ol hydrochloride,3r-piperidin-3-ol hydrochloride,r-+-3-hydroxypiperidine hydrochloride,r-3-hydroxypiperidine hydrocloride,r-3-hydroxypiperidine hcl,r-+-3-piperidinol hydrochloride,r-3-hydroxylpiperidine hydrochloride,3r-3-piperidinol hydrochloride,r-3-piperidinol hydrochloride
2759333
(3R)-piperidin-3-ol;hydrochloride
C1CC(CNC1)O.Cl
198976-43-1
C5H12ClNO
137.61
MFCD00192221
VLECDMDGMKPUSK-NUBCRITNSA-N
r-3-hydroxypiperidine hydrochloride,r-piperidin-3-ol hydrochloride,3r-piperidin-3-ol hydrochloride,r-+-3-hydroxypiperidine hydrochloride,r-3-hydroxypiperidine hydrocloride,r-3-hydroxypiperidine hcl,r-+-3-piperidinol hydrochloride,r-3-hydroxylpiperidine hydrochloride,3r-3-piperidinol hydrochloride,r-3-piperidinol hydrochloride
2759333
(3R)-piperidin-3-ol;hydrochloride
C1CC(CNC1)O.Cl
Melting Point 134°C to 138°C
Specific Rotation + 7.1
Color White
Quantity 1g
Elemental Analysis 42.18 to 44.73%, 9.92 to 10.43%
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 137.61
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material (R)-(+)-3-Hydroxypiperidine hydrochloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning