Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Phenylcyclopentanone 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
1g
86505-44-4
C11H12O
160.216
MFBXYJLOYZMFIN-SNVBAGLBSA-N
11030117
(3R)-3-phenylcyclopentan-1-one
C1CC(=O)CC1C2=CC=CC=C2
86505-44-4
C11H12O
160.216
MFBXYJLOYZMFIN-SNVBAGLBSA-N
11030117
(3R)-3-phenylcyclopentan-1-one
C1CC(=O)CC1C2=CC=CC=C2
Color Yellow
Quantity 1g
Formula Weight 160.22
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material (R)-3-Phenylcyclopentanone