Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Pyrrolidinol hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
104706-47-0
C4H9NO·ClH
123.58
QPMSJEFZULFYTB-PGMHMLKASA-N
r-3-hydroxypyrrolidine hydrochloride,r---3-pyrrolidinol hydrochloride,3r-pyrrolidin-3-ol hydrochloride,r-pyrrolidin-3-ol hydrochloride,r-3-hydroxypyrrolidine hcl,r-3-pyrrolidinol hydrochloride,3r-3-hydroxypyrrolidine hydrochloride,3r-pyrrolidinol hydrochloride,r-+-3-pyrrolidinol hydrochloride
2759336
(3R)-pyrrolidin-3-ol;hydrochloride
C1CNCC1O.Cl
104706-47-0
C4H9NO·ClH
123.58
QPMSJEFZULFYTB-PGMHMLKASA-N
r-3-hydroxypyrrolidine hydrochloride,r---3-pyrrolidinol hydrochloride,3r-pyrrolidin-3-ol hydrochloride,r-pyrrolidin-3-ol hydrochloride,r-3-hydroxypyrrolidine hcl,r-3-pyrrolidinol hydrochloride,3r-3-hydroxypyrrolidine hydrochloride,3r-pyrrolidinol hydrochloride,r-+-3-pyrrolidinol hydrochloride
2759336
(3R)-pyrrolidin-3-ol;hydrochloride
C1CNCC1O.Cl
Melting Point 102°C to 107.5°C
Packaging Glass Bottle
Specific Rotation -7.6
Quantity 10g
Assay Percent Range 97.5% min. (Argentometry)
Formula Weight 123.58
Percent Purity 98%
Chemical Name or Material (R)-3-Pyrrolidinol hydrochloride

GHS Signal Word: Warning