Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI™ R-300 Rotavapor™ Electronic Lift Available on GSA/VA Contract for Federal Government customers only.

The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor™ R-300 to a fully integrated system where a central interface regulates each component.

$7450.00 - $7900.00

Specifications

Capacity (Metric) 5L
Certifications CE / CSA
Hertz 50 to 60 Hz
Model R-300
Product Type Rotary Evaporator
View More Specs

Products 2
Catalog Number Mfr. No. Condenser Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Condenser Type For Use With (Application) Price Quantity & Availability  
05-000-947 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
11R300252C102
Cold Trap Condenser Distillation of Low Boiling-Point Solvents (for use with dry ice) Each for $7,900.00
Add to cart
 
05-000-977 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
11R300252V102
Vertical Condenser Standard Distillation (for use with cold water or a chiller) Each for $7,450.00
Add to cart
 
Description

Description

  • Intuitive user interaction
  • Precise operation control
  • 5L flask capacity
  • Ergonomic handle with electronic lift
  • Combi-Clip
  • Additional glass assemblies and configurations

Certifications

CE and CSA certified

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

5L
50 to 60 Hz
Rotary Evaporator
24/40
100 to 120V
CE / CSA
R-300
10 to 280 rpm
20 to 220°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit