Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-4-bromo-α-methylbenzyl alcohol, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
76155-78-7
C8H9BrO
201.063
MFCD06201861
XTDTYSBVMBQIBT-ZCFIWIBFSA-N
r-1-4-bromophenyl ethanol,1r-1-4-bromophenyl ethanol,1r-1-4-bromophenyl ethan-1-ol,r-4-bromo-alpha-methylbenzyl alcohol,r-4-bromo-alpha-methylbenzylalcohol,pubchem14252,r-4'-bromo-1-phenylethanol,r-1-4-bromophenyl ethanol; r-4 inverted exclamation marka-bromo-1-phenylethanol
6950173
(1R)-1-(4-bromophenyl)ethanol
CC(C1=CC=C(C=C1)Br)O
76155-78-7
C8H9BrO
201.063
MFCD06201861
XTDTYSBVMBQIBT-ZCFIWIBFSA-N
r-1-4-bromophenyl ethanol,1r-1-4-bromophenyl ethanol,1r-1-4-bromophenyl ethan-1-ol,r-4-bromo-alpha-methylbenzyl alcohol,r-4-bromo-alpha-methylbenzylalcohol,pubchem14252,r-4'-bromo-1-phenylethanol,r-1-4-bromophenyl ethanol; r-4 inverted exclamation marka-bromo-1-phenylethanol
6950173
(1R)-1-(4-bromophenyl)ethanol
CC(C1=CC=C(C=C1)Br)O
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Color Colorless to Yellow
Quantity 1g
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3CH(OH)C6H4Br
Formula Weight 201.06
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material (R)-4-Bromo-α-methylbenzyl alcohol
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.