Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-4-Fluoro-alpha-methylbenzyl alcohol, 95% (98% E.E.), Acros Organics

Click to view available options
Quantity:
250mg
Packaging:
Glass Bottle
101219-68-5
C8H9FO
140.16
MFCD03092997
PSDSORRYQPTKSV-ZCFIWIBFSA-N
r-1-4-fluorophenyl ethanol,1r-1-4-fluorophenyl ethanol,1r-1-4-fluorophenyl ethan-1-ol,r-4-fluoro-alpha-methylbenzyl alcohol,pubchem14245,r-4-fluorophenylethan-1-ol,r-p-fluoro-alpha-methylbenzylalcohol,r-+-1-4-fluorophenyl ethanol,r-4-fluoro-,a-methylbenzyl alcohol,benzenemethanol,4-fluoro-a-methyl-, ar
852998
(1R)-1-(4-fluorophenyl)ethanol
CC(C1=CC=C(C=C1)F)O
101219-68-5
C8H9FO
140.16
MFCD03092997
PSDSORRYQPTKSV-ZCFIWIBFSA-N
r-1-4-fluorophenyl ethanol,1r-1-4-fluorophenyl ethanol,1r-1-4-fluorophenyl ethan-1-ol,r-4-fluoro-alpha-methylbenzyl alcohol,pubchem14245,r-4-fluorophenylethan-1-ol,r-p-fluoro-alpha-methylbenzylalcohol,r-+-1-4-fluorophenyl ethanol,r-4-fluoro-,a-methylbenzyl alcohol,benzenemethanol,4-fluoro-a-methyl-, ar
852998
(1R)-1-(4-fluorophenyl)ethanol
CC(C1=CC=C(C=C1)F)O
Packaging Glass Bottle
Color Colorless to Yellow
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Quantity 250mg
Linear Formula CH3CH(OH)C6H4F
Formula Weight 140.16
Physical Form Liquid
Percent Purity 95% (98% E.E.)
Chemical Name or Material (R)-(+)-4-Fluoro-α-methylbenzyl alcohol

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning