Alfa Aesar™ (R)-(-)-4-Fluorostyrene oxide, 98+%, ee 98+%

Manufacturer: Alfa Aesar™ H3360303

Quantity 1g

 View more versions of this product

Catalog No. AAH3360303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 134356-73-3
    Molecular Formula C8H7FO
    Formula Weight 138.14
    MDL Number MFCD03788740
    UN Number UN1992
    Synonym (R)-(-)-2-(4-Fluorophenyl)oxirane
    Chemical Name or Material (R)-(-)-4-Fluorostyrene oxide

    Specifications

    Formula Weight 138.14
    Molecular Formula C8H7FO
    MDL Number MFCD03788740
    UN Number UN1992
    Synonym (R)-(-)-2-(4-Fluorophenyl)oxirane
    Boiling Point 92°C (14mmHg)
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H331-H226-H351-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Flammable liquid and vapor.
    Suspected of causing cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    CAS 134356-73-3
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material (R)-(-)-4-Fluorostyrene oxide
    Assay ≥98%, ee ≥98%

    Description

    Applications
    It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

    Solubility
    Sparingly soluble in water.(0.26 g/L) (25°C),

    Notes
    Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

    GHS H Statement
    H331-H226-H351-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Flammable liquid and vapor.
    Suspected of causing cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit