Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-4-Hydroxy-2-pyrrolidinone, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
22677-21-0
C4H7NO2
101.11
IOGISYQVOGVIEU-GSVOUGTGSA-N
r-4-hydroxypyrrolidin-2-one,r-+-4-hydroxy-2-pyrrolidinone,r-4-hydroxy-2-pyrrolidone,4r-4-hydroxypyrrolidin-2-one,r-4-hydroxy-2-pyrrolidinone,4r-4-hydroxy-2-pyrrolidinone,r-4-hydroxy-pyrrolidin-2-one,2-pyrrolidinone, 4-hydroxy-, 4r,r-+-4-hydroxy-2-pyrrolidone,rhpd
185505
(4R)-4-hydroxypyrrolidin-2-one
C1C(CNC1=O)O
22677-21-0
C4H7NO2
101.11
IOGISYQVOGVIEU-GSVOUGTGSA-N
r-4-hydroxypyrrolidin-2-one,r-+-4-hydroxy-2-pyrrolidinone,r-4-hydroxy-2-pyrrolidone,4r-4-hydroxypyrrolidin-2-one,r-4-hydroxy-2-pyrrolidinone,4r-4-hydroxy-2-pyrrolidinone,r-4-hydroxy-pyrrolidin-2-one,2-pyrrolidinone, 4-hydroxy-, 4r,r-+-4-hydroxy-2-pyrrolidone,rhpd
185505
(4R)-4-hydroxypyrrolidin-2-one
C1C(CNC1=O)O
Melting Point 155°C to 156°C
Specific Rotation 59.4
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 101.11
Percent Purity 97%
Chemical Name or Material (R)-(+)-4-Hydroxy-2-pyrrolidinone

GHS Signal Word: Warning