Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-[(5,6),(5',6')-bis(ethylenedioxy)biphenyl-2,2'-diyl](diphenylphosphine), 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
250mg
Packaging:
Glass bottle
445467-61-8
C40H32O4P2
638.64
MFCD05861608
GDMCOFXEPNHXJT-UHFFFAOYSA-N
synphos,s-synphos,s-6,6'-bis diphenylphosphino-2,2',3,3'-tetrahydro-5,5'-bibenzo b 1,4 dioxine,r-+-6,6'-bis diphenylphosphino-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin,r-synphos,s-synphos tm,ar-2,2',3,3'-tetrahydro-6,6'-bis diphenylphosphino-5,5'-bi 1,4-benzodioxin,6,6'-bis diphenylphosphino-2,2',3,3'-tetrahydro-5,5'-bibenzo b 1,4 dioxine,r-+-6,6-bis diphenylphosphino-2,2,3,3-tetrahydro-5,5-bi-1,4-benzodioxin,5-6-diphenylphosphanyl-2,3-dihydro-1,4-benzodioxin-5-yl-2,3-dihydro-1,4-benzodioxin-6-yl diphenylphosphane
11193918
[5-(6-diphenylphosphanyl-2,3-dihydro-1,4-benzodioxin-5-yl)-2,3-dihydro-1,4-benzodioxin-6-yl]-diphenylphosphane
C1COC2=C(O1)C=CC(=C2C3=C(C=CC4=C3OCCO4)P(C5=CC=CC=C5)C6=CC=CC=C6)P(C7=CC=CC=C7)C8=CC=CC=C8
445467-61-8
C40H32O4P2
638.64
MFCD05861608
GDMCOFXEPNHXJT-UHFFFAOYSA-N
synphos,s-synphos,s-6,6'-bis diphenylphosphino-2,2',3,3'-tetrahydro-5,5'-bibenzo b 1,4 dioxine,r-+-6,6'-bis diphenylphosphino-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin,r-synphos,s-synphos tm,ar-2,2',3,3'-tetrahydro-6,6'-bis diphenylphosphino-5,5'-bi 1,4-benzodioxin,6,6'-bis diphenylphosphino-2,2',3,3'-tetrahydro-5,5'-bibenzo b 1,4 dioxine,r-+-6,6-bis diphenylphosphino-2,2,3,3-tetrahydro-5,5-bi-1,4-benzodioxin,5-6-diphenylphosphanyl-2,3-dihydro-1,4-benzodioxin-5-yl-2,3-dihydro-1,4-benzodioxin-6-yl diphenylphosphane
11193918
[5-(6-diphenylphosphanyl-2,3-dihydro-1,4-benzodioxin-5-yl)-2,3-dihydro-1,4-benzodioxin-6-yl]-diphenylphosphane
C1COC2=C(O1)C=CC(=C2C3=C(C=CC4=C3OCCO4)P(C5=CC=CC=C5)C6=CC=CC=C6)P(C7=CC=CC=C7)C8=CC=CC=C8
Packaging Glass bottle
Quantity 100mg
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 638.64
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material (R)-(+)-[(5,6),(5';,6';)-Bis(ethylenedioxy)biphenyl-2,2';-diyl](diphenylphosphine)
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning