Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene 95.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
911200-98-1
C10H14O
150.221
WBRYUDKSKFXYOD-JTQLQIEISA-N
14942946
(3aR)-3a-prop-2-enyl-2,3,4,5-tetrahydrocyclopenta[b]furan
C=CCC12CCC=C1OCC2
911200-98-1
C10H14O
150.221
WBRYUDKSKFXYOD-JTQLQIEISA-N
14942946
(3aR)-3a-prop-2-enyl-2,3,4,5-tetrahydrocyclopenta[b]furan
C=CCC12CCC=C1OCC2
Color Yellow
Quantity 1g
Formula Weight 150.22
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material (R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene