Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-5-Hydroxymethyl-2-pyrrolidinone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
66673-40-3
C5H9NO2
115.13
MFCD00077791
HOBJEFOCIRXQKH-SCSAIBSYSA-N
r---5-hydroxymethyl-2-pyrrolidinone,d-pyroglutaminol,5r-5-hydroxymethyl pyrrolidin-2-one,r-5-hydroxymethyl pyrrolidin-2-one,r-5-hydroxymethyl-2-pyrrolidinone,r-5-hydroxymethyl-pyrrolidin-2-one,--d-pyroglutamol,r---5-hydroxymethylpyrrolidin-2-one,5-hydroxymethyl-2-pyrrolidinone
7128291
(5R)-5-(hydroxymethyl)pyrrolidin-2-one
C1CC(=O)NC1CO
66673-40-3
C5H9NO2
115.13
MFCD00077791
HOBJEFOCIRXQKH-SCSAIBSYSA-N
r---5-hydroxymethyl-2-pyrrolidinone,d-pyroglutaminol,5r-5-hydroxymethyl pyrrolidin-2-one,r-5-hydroxymethyl pyrrolidin-2-one,r-5-hydroxymethyl-2-pyrrolidinone,r-5-hydroxymethyl-pyrrolidin-2-one,--d-pyroglutamol,r---5-hydroxymethylpyrrolidin-2-one,5-hydroxymethyl-2-pyrrolidinone
7128291
(5R)-5-(hydroxymethyl)pyrrolidin-2-one
C1CC(=O)NC1CO
Quantity 5g
Color White to Yellow
Boiling Point 147.0°C to 149.0°C
Melting Point 77.0°C to 84.0°C
Specific Rotation Condition −31.00 (20.00°C c=1,C2H5OH)
Specific Rotation − 31.00
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 115.13
Physical Form Crystalline Powder or Flakes
Percent Purity 98%
Chemical Name or Material (R)-(-)-5-Hydroxymethyl-2-pyrrolidinone, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA