Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-8-(Diphenylphosphino)-1,2,3,4-tetrahydronaphthalen-1-amine, 97%, Thermo Scientific™

Catalog Number ac434112500
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
960128-64-7
C22H22NP
331.40
MFCD16621442
KTWOFTBMHDQUEE-GNLPSFAGNA-N
r-8-diphenylphosphino-1,2,3,4-tetrahydronaphthalen-1-amine, r-1-amino-8-diphenylphosphino-1,2,3,4-tetrahydronaphthalene, 1r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine, 1r-8-diphenylphosphino tetralin-1-amine, 1r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-ami, r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine, r---8-diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, r---8-diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, 97+%
71309688
(1R)-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine
N[C@@H]1CCCC2=C1C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
960128-64-7
C22H22NP
331.40
MFCD16621442
KTWOFTBMHDQUEE-GNLPSFAGNA-N
r-8-diphenylphosphino-1,2,3,4-tetrahydronaphthalen-1-amine, r-1-amino-8-diphenylphosphino-1,2,3,4-tetrahydronaphthalene, 1r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine, 1r-8-diphenylphosphino tetralin-1-amine, 1r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-ami, r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine, r---8-diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, r---8-diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, 97+%
71309688
(1R)-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine
N[C@@H]1CCCC2=C1C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 127.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 331.39
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (R)-8-(Diphenylphosphino)-1,2,3,4-tetrahydronaphthalen-1-amine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit