Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-α-Allylalanine hydrate, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
1266371-17-8,96886-56-5
C6H11NO2·xH2O
129.16
(R)-(+)-2-Amino-2-methyl-4-pentenoic acid hydrate
1266371-17-8,96886-56-5
C6H11NO2·xH2O
129.16
(R)-(+)-2-Amino-2-methyl-4-pentenoic acid hydrate
Melting Point 271°C to 276°C
Specific Rotation 26
Color White to Yellow
Enantiomeric Excess 98.0% min.
Quantity 100mg
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 to 102.5% (as Monohydrate)
Formula Weight 129.16
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (R)-α-Allylalanine hydrate, 98% ee
Show More Show Less