Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol, 98%, Acros Organics

Click to view available options
Quantity:
250mg
22348-32-9
C17H19NO
253.345
MFCD00077754
OGCGXUGBDJGFFY-MRXNPFEDSA-N
r-diphenyl pyrrolidin-2-yl methanol,r-+-alpha,alpha-diphenyl-2-pyrrolidinemethanol,r-+-2-diphenylhydroxymethyl pyrrolidine,diphenyl 2r-pyrrolidin-2-ylmethanol,r-+-2-diphenylhydroxymethyl-pyrrolidin,diphenyl-2r-pyrrolidin-2-yl methanol,r-alpha,alpha-diphenyl-2-pyrrolidinemethanol,alpha,alpha-diphenyl-d-prolinol,r-+-alpha,alpha-diphenylprolinol,diphenyl 2r-pyrrolidin-2-yl methanol
7045371
diphenyl-[(2R)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O
22348-32-9
C17H19NO
253.345
MFCD00077754
OGCGXUGBDJGFFY-MRXNPFEDSA-N
r-diphenyl pyrrolidin-2-yl methanol,r-+-alpha,alpha-diphenyl-2-pyrrolidinemethanol,r-+-2-diphenylhydroxymethyl pyrrolidine,diphenyl 2r-pyrrolidin-2-ylmethanol,r-+-2-diphenylhydroxymethyl-pyrrolidin,diphenyl-2r-pyrrolidin-2-yl methanol,r-alpha,alpha-diphenyl-2-pyrrolidinemethanol,alpha,alpha-diphenyl-d-prolinol,r-+-alpha,alpha-diphenylprolinol,diphenyl 2r-pyrrolidin-2-yl methanol
7045371
diphenyl-[(2R)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O
Specific Rotation Condition + 59.00 (25.00°C 589nm,c=3,MeOH)
Infrared Spectrum Authentic
Color White to Yellow
Specific Rotation +57° to +61° (25°C, 589nm) (c=3, MeOH)
Quantity 250mg
Melting Point 77°C to 81°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 253.34
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material (R)-(+)-α, α-Diphenyl-2-pyrrolidinemethanol, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA