Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-alpha,alpha-Diphenylprolinol, 99%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
22348-32-9
C17H19NO
253.345
MFCD00077754
OGCGXUGBDJGFFY-MRXNPFEDSA-N
r-diphenyl pyrrolidin-2-yl methanol, r-+-alpha,alpha-diphenyl-2-pyrrolidinemethanol, r-+-2-diphenylhydroxymethyl pyrrolidine, diphenyl 2r-pyrrolidin-2-ylmethanol, r-+-2-diphenylhydroxymethyl-pyrrolidin, diphenyl-2r-pyrrolidin-2-yl methanol, r-alpha,alpha-diphenyl-2-pyrrolidinemethanol, alpha,alpha-diphenyl-d-prolinol, r-+-alpha,alpha-diphenylprolinol, diphenyl 2r-pyrrolidin-2-yl methanol
7045371
diphenyl-[(2R)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O
22348-32-9
C17H19NO
253.345
MFCD00077754
OGCGXUGBDJGFFY-MRXNPFEDSA-N
r-diphenyl pyrrolidin-2-yl methanol, r-+-alpha,alpha-diphenyl-2-pyrrolidinemethanol, r-+-2-diphenylhydroxymethyl pyrrolidine, diphenyl 2r-pyrrolidin-2-ylmethanol, r-+-2-diphenylhydroxymethyl-pyrrolidin, diphenyl-2r-pyrrolidin-2-yl methanol, r-alpha,alpha-diphenyl-2-pyrrolidinemethanol, alpha,alpha-diphenyl-d-prolinol, r-+-alpha,alpha-diphenylprolinol, diphenyl 2r-pyrrolidin-2-yl methanol
7045371
diphenyl-[(2R)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 76°C to 78°C
Quantity 250mg
Optical Rotation +57.5° (c=3 in Methanol)
Beilstein 4353363
Formula Weight 253.35
Percent Purity 99%
Chemical Name or Material (R)-(+)-alpha,alpha-Diphenylprolinol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only