Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride monohydrate, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
104347-13-9
C4H7NO2·ClH
155.59
XBKCXPRYTLOQKS-AENDTGMFSA-N
d-homoserine lactone hydrochloride,r-+-alpha-amino-gamma-butyrolactone hydrochloride,r-3-aminodihydrofuran-2 3h-one hydrochloride,3r-3-aminotetrahydrofuran-2-one hydrochloride,homoserine lactone hydrochloride,2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r,r-+-a-amino-g-butyrolactone hydrochloride,pubchem14743,r-+-,a-amino-,a-butyrolactone hydrochloride,d-homoserine lactone hcl
11954435
(3R)-3-aminooxolan-2-one;hydrochloride
C1COC(=O)C1N.Cl
104347-13-9
C4H7NO2·ClH
155.59
XBKCXPRYTLOQKS-AENDTGMFSA-N
d-homoserine lactone hydrochloride,r-+-alpha-amino-gamma-butyrolactone hydrochloride,r-3-aminodihydrofuran-2 3h-one hydrochloride,3r-3-aminotetrahydrofuran-2-one hydrochloride,homoserine lactone hydrochloride,2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r,r-+-a-amino-g-butyrolactone hydrochloride,pubchem14743,r-+-,a-amino-,a-butyrolactone hydrochloride,d-homoserine lactone hcl
11954435
(3R)-3-aminooxolan-2-one;hydrochloride
C1COC(=O)C1N.Cl
Specific Rotation 30
Melting Point 215.1°C to 217.1°C
Color Yellow to Brown
Packaging Glass bottle
Quantity 1g
Water 15% max.
Infrared Spectrum Authentic
Elemental Analysis 29.64 to 32.12% C, 8.64 to 9.36% N
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Formula Weight 155.59
Physical Form Powder, Solid or Crystalline Powder
Percent Purity 97%
Chemical Name or Material (R)-(+)-α-Amino-γ-butyrolactone hydrochloride monohydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning