Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Baclofen, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

69308-37-8
C10H12ClNO2
213.661
KPYSYYIEGFHWSV-QMMMGPOBSA-N
(R)-4-Amino-3-(4-chlorophenyl)butanoic acid
44602

Selective GABAB agonist. Active enantiomer of (RS)-Baclofen (Cat. No. 0417)

More active enantiomer of (RS)-Baclofen (Cat. No. 0417), a selective GABAB agonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

SMILES C1=CC(=CC=C1C(CC(=O)O)CN)Cl
IUPAC Name (3R)-4-amino-3-(4-chlorophenyl)butanoic acid
Formula Weight 213.66
Quantity 10mg
Chemical Name or Material (R)-Baclofen
Grade >99%

Recommended Storage : Store at Room Temperature