Tocris™ (R)-CR8

Dual cdk1/cdk5 inhibitor; also inhibits CK1

Manufacturer: Tocris™ 3605/50

 View more versions of this product

Catalog No. 36-055-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors (R)-CR8
    Purity >99
    Molecular Weight 540.92
    Quantity 50mg
    Product Type (R)-CR8

    Inhibitor of cyclin-dependent kinase (cdk) and casein kinase 1 (CK1) (IC50 values are 0.13µM for cdk1/cyclin B and cdk5/p25; and 0.6µM for CK1).

    • Chemical Name: 2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-(4-(2-pyridyl)benzyl)-9-isopropylpurine trihydrochloride
    • Formula: C24H29N7O.3HCl
    • CAS Number: 294646-77-8
    • Solubility: Soluble to 100mM in water and to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit