Learn More
R&D Systems™ 6-Biotin-17-NAD
Provides a non-isotopic alternative to radiolabeled NAD for assays requiring this substrate.
Manufacturer:  R&D Systems™ 467050001
Catalog No. 467050001
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Biotinylated NAD (6-Biotin-17-NAD) provides a non-isotopic alternative to radiolabeled NAD for assays requiring this substrate. For instance, biotinylated NAD can be used as an indirect measure of PARP activity when biotin incorporation is detected using a streptavidin detection system.
Specifications
Specifications
Store the unopened product at -80°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.