Learn More
Tocris Bioscience™ ABT 089 Dihydrochloride
High affinity and selective α4β2 nAChR partial agonist
$179.60 - $752.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
507910R
View Documents
Tocris Bioscience™
5079/10
10 mg
Each for $179.60
Add to cart
507950R
View Documents
Tocris Bioscience™
5079/50
50 mg
Each for $752.00
Add to cart
Description
Description
High affinity and selective α4β2 partial agonist (Ki = 16 nM). Exhibits >1000-fold and >10,000-fold selectivity for α4β2 over α1β1γ1 and α7 receptors respectively. Neuroprotective against glutamate-induced toxicity in rat cortical neurons in vitro. Enhances cognitive performance in vivo. Orally bioavailable and brain penetrant.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use