Learn More
Tocris Bioscience™ AM 6545
High affinity and selective CB1 antagonist
$248.00 - $1027.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
544310
View Documents
Tocris Bioscience™
5443/10
10 mg
Each for $248.00
Add to cart
544350
View Documents
Tocris Bioscience™
5443/50
50 mg
Each for $1,027.00
Add to cart
Description
Description
High affinity and selective CB1 antagonist (Ki = 3.3 nM). Exhibits >100-fold selectivity for CB1 over CB2 receptors. Improves glucose homeostasis, reverses hepatic steatosis and reduces body weight in diet-induced obese mice. Orally available and non-brain penetrant.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use