Learn More
Tocris Bioscience™ AZ 191
Potent DYRK1B inhibitor
$209.00 - $878.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
523210
View Documents
Tocris Bioscience™
5232/10
10 mg
Each for $209.00
Add to cart
523250
View Documents
Tocris Bioscience™
5232/50
50 mg
Each for $878.00
Add to cart
Description
Description
Potent DYRK1B inhibitor (IC50 values are 17, 88 and 1890 nM for DYRK1B, DYRK1A and DYRK2 respectively). Inhibits DYRK1B-induced Gi phase cell cycle arrest in Panc-1 cells in vitro.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use