R&D Systems BRD 7116, Tocris™

Inhibitor of leukemia stem cell (LSC) activity

Manufacturer: R&D Systems 5336/50

 View all options for this product

Catalog No. 533650

  Check Availability
Add to Cart


Research Use Only. Not For Human or Veterinary Use


    Chemical Name or Material BRD 7116
    CAS 329059-55-4
    Assay Percent Range >98%
    Molecular Formula C28H36N2O4S
    Formula Weight 496.66
    Synonym N,N'-(Sulfonyldi-4,1-phenylene)bis[2,2,3,3-tetramethylcyclopropanecarboxamide]
    Recommended Storage Store at +4°C.
    Quantity 50mg
    Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
    Physical Form Solid
    Product Type Bioactive Small Molecules

    • Exhibits >100-fold selectivity for LSCs over normal hematopoietic stem cells

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit