Learn More
Tocris Bioscience™ BRD 7116
Inhibitor of leukemia stem cell activity
$95.00 - $399.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
533610 View Documents Tocris Bioscience™
5336/10
Each for $95.00
Add to cart
533650 View Documents Tocris Bioscience™
5336/50
Each for $399.00
Add to cart
Description
Description
Inhibitor of leukemia stem cell (LSC) activity (EC50 = 200 nM). Exhibits >100-fold selectivity for LSCs over normal hematopoietic stem cells.
Research Use Only. Not For Human or Veterinary Use