Learn More
Tocris Bioscience™ C 87
TNF-α inhibitor
$181.00 - $760.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
548410
View Documents
Tocris Bioscience™
5484/10
10 mg
Each for $181.00
Add to cart
548450
View Documents
Tocris Bioscience™
5484/50
50 mg
Each for $760.00
Add to cart
Description
Description
TNF-α inhibitor (Kd = 110 nM for hTNF-α). Inhibits TNF-α-induced cytotoxicity in L929 fibroblast cells. Attenuates inflammation and increases survival in a mouse hepatitis model.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use