Learn More
R&D Systems™ Calcein AM Cell Viability Assay Available on GSA/VA Contract for Federal Government customers only.
Provides a simple, rapid, and accurate method to measure cell viability and/or cytotoxicity
Manufacturer:  R&D Systems™ 4892010K
Catalog No. 4892010K
Add to cart
This item is not returnable. View return policy
Description
Description
Suitable for proliferating and non-proliferating cells, for both suspension and adherent cells
Non-radioactive microplate
No solubilization step as in an MTT assay
Well-suited for high-throughput assays
Excellent retention and brightness compared to o
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1000 Tests
The product requires storage at -20 to -70° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.