Learn More
R&D Systems™ Calf Thymus DNA Available on GSA/VA Contract for Federal Government customers only.
5 vials (1 mL/vial) of Calf Thymus DNA
Manufacturer:  R&D Systems™ 96005D
Catalog No. 96005D
Add to cart
This item is not returnable. View return policy
Description
Description
  • Fragment size range of 200 - 500bp
  • The format allows convenient addition to buffers to create a final recommended working concentration of 0.1mg/mL
  • Quality control testing includes concentration determination, fragment sizing by gel electrophoresis, A260/A280 ultraviolet absorbance ratio, and protein content
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
10mg/mL
Calf Thymus DNA
Store at -20° C in a manual defrost freezer. For long-term storage, freeze in working aliquots at -80° C. Avoid freeze-thaw cycles.
Highly purified and qualified for Northern, Southern, probe array, and dot blotting procedures,
5 x 1mL