Learn More
R&D Systems™ CometAssay™ High Throughput Slides
Provides convenient, simple and reliable sample preparation, processing, analysis and storage
Supplier:  R&D Systems™ 425202K01
Catalog No. 425202K01
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Features a writing surface, specially treated glass surface and hydrophobic barrier for sample
3 x 2 in. (75 x 50mm)
Specifications
Specifications
100 Slides
Product Suggestions
Product Suggestions