Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Cotton Rat CCL4/MIP-1 beta Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$487.20
Specifications
Molecular Weight (g/mol) | 7.8 kDa |
---|---|
Name | CCL4/MIP-1 beta |
Quantity | 25μg |
Source | E. coli-derived cotton rat CCL4/MIP-1 beta protein Ala24-Asn92 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
1024MB025
|
R&D Systems™
1024MB025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
Each for $487.20
|
|
|||||
1024MB025CF
|
R&D Systems™
1024MB025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
Each for $487.20
|
|
|||||
Description
The Recombinant Cotton Rat CCL4/MIP-1 beta Protein is derived from E. coli. The Recombinant Cotton Rat CCL4/MIP-1 beta Protein has been validated for the following applications: Bioactivity.Specifications
7.8 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
CCL4/MIP-1 beta | |
E. coli-derived cotton rat CCL4/MIP-1 beta protein Ala24-Asn92 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title