Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Cotton Rat IL-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$473.28
Specifications
Molecular Weight (g/mol) | 15 kDa |
---|---|
Name | IL-4 |
Quantity | 10μg |
Source | E. coli-derived cotton rat IL-4 protein Cys21-Phe147,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
584R4010
|
R&D Systems™
584R4010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $473.28
|
|
|||||
584R4010CF
|
R&D Systems™
584R4010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
Each for $473.28
|
|
|||||
Description
The Recombinant Cotton Rat IL-4 Protein is derived from E. coli. The Recombinant Cotton Rat IL-4 Protein has been validated for the following applications: Bioactivity.Specifications
15 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
IL-4 | |
E. coli-derived cotton rat IL-4 protein Cys21-Phe147,with an N-terminal Met | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title