Learn More
Tocris Bioscience™ CU-T12-9
Potent TLR1/2 agonist
$222.00 - $927.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
541410
View Documents
Tocris Bioscience™
5414/10
10 mg
Each for $222.00
Add to cart
541450
View Documents
Tocris Bioscience™
5414/50
50 mg
Each for $927.00
Add to cart
Description
Description
Potent TLR1/2 agonist (EC50 = 52.9 nM). Activates the NF-κB pathway and upregulates proinflammatory cytokines in vitro. Also enhances TLR1 and TLR2 dimerization.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use