R&D Systems CultreCoat™ BME Cell Invasion Assays, 96 well
Reagent kit for evaluating cell invasion for low, intermediary or highly invasive cell models
Manufacturer: R&D Systems 3483096K
 View more versions of this product
Catalog No. 3483096K
  Check Availability
Add to Cart
    Specifications
    Includes 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50μg
    Quantity 1 Kit
    Type High BME Cell Invasion Assay
  • Description & Specifications
Related Products
Product Content Feedback