Learn More
R&D Systems™ Cultrex™ BME Cell Invasion Assay Available on GSA/VA Contract for Federal Government customers only.
Is a 96-well microplate-based assay designed to accelerate the screening process for compounds that influence chemotaxis
Manufacturer:  R&D Systems™ 3455096K
Catalog No. 3455096K
Add to cart
Includes: Cell Invasion Chamber; BME Coating Solution; Coating Buffer; Cell Wash Buffer; Cell Dissociation Solution; Calcein AM
This item is not returnable. View return policy
Description
Description
Invasive migration is a fundamental function underlying cellular processes such as angiogenesis, embryonic development, immune response, metastasis, and invasion of cancer cells.
Flexible, standardized, high-throughput format for quantitating the degree to which invasive cells penetrate a barrier consisting of Basement Membrane Extract (BME) in response to chemoattractants and/or inhibiting compounds
This assay kit contains all t
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Each
The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.