Learn More
R&D Systems™ Cultrex™ Collagen I Cell Invasion Assay
Reagent kit for investigating chemotaxis, cell migration, and/or cell invasion
$359.00 - $389.00
Specifications
Type Cultrex Collagen I Cell Invasion Assay
Includes 5X Collagen I Solution, 10X Coating Buffer, 25X Cell Wash Buffer, 10X Cell Dissociation Solution, Calcein-AM
View More Specs
Includes: 96 Well Cell Invasion Chamber; 5X Collagen I Solution, 1mL; 10X Coating Buffer, 1mL; 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50ug
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
3457096K
View Documents
R&D Systems™
3457096K
Each
Each for $359.00
Add to cart
3457024K
View Documents
R&D Systems™
3457024K
24 Inserts per Pack
Each for $389.00
Add to cart
Description
Description
Collagens comprise a large family of insoluble extracellular glycoproteins that are essential components of connective tissues such as tendons, ligaments, cartilage, bone and skin. The mature polypeptides are secreted as coiled, left-handed helices that s
Flexible, standardized, high-throughput format for quantitating the degree to which invasive cells penetrate a barrier consisting of extracellular matrix protein Collagen I in response to chemoattractants and/or inhibiting compounds
Features a simplified
Specifications
Specifications
Cultrex Collagen I Cell Invasion Assay
5X Collagen I Solution, 10X Coating Buffer, 25X Cell Wash Buffer, 10X Cell Dissociation Solution, Calcein-AM