R&D Systems Cultrex™ Collagen I Cell Invasion Assay
Reagent kit for investigating chemotaxis, cell migration, and/or cell invasion
Manufacturer: R&D Systems 3457024K
 View more versions of this product
Catalog No. 3457024K
  Check Availability
Add to Cart
    Specifications
    Includes 5X Collagen I Solution, 10X Coating Buffer, 25X Cell Wash Buffer, 10X Cell Dissociation Solution, Calcein-AM
    Quantity 24 Inserts per Pack
    Type Cultrex Collagen I Cell Invasion Assay
  • Description & Specifications
Related Products
Product Content Feedback